Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252250.1 | NP_001239179.1 | 16 - 17 | 2349 | 74 | |
GenBank mRNA | L22988.1 | - | 16 - 17 | 1807 | 74 | |
L27105.1 | - | 16 - 17 | 1800 | 74 | ||
L28838.1 | - | 2 - 3 | 72 | 74 | ||
X75759.1 | - | 5 - 6 | 593 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |