Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039176.2 | NP_001034265.1 | 1 - 2 | 129 | 68 | |
GenBank mRNA | AK156003.1 | - | 1 - 2 | 1723 | 68 | |
BC006735.1 | - | 1 - 2 | 129 | 3156004 | 68 | |
BY773043.1 | - | 1 - 2 | 83 | 68 | ||
CJ094566.1 | - | 1 - 2 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |