Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173719.3 | NP_776067.1 | 7 - 8 | 687 | 93 | |
GenBank mRNA | AK005422.1 | - | 7 - 8 | 670 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons |