Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168289.1 | NP_001161761.1 | 2 - 3 | 707 | 87 | |
NM_027772.2 | NP_082048.2 | 2 - 3 | 707 | 87 | ||
GenBank mRNA | AB210840.1 | - | 2 - 3 | 431 | 87 | |
AK017329.1 | - | 2 - 3 | 677 | 87 | ||
AK140091.1 | - | 2 - 3 | 594 | 87 | ||
AK165732.1 | - | 2 - 3 | 706 | 87 | ||
BC147693.1 | - | 2 - 3 | 484 | 9056666 | 87 | |
BC147694.1 | - | 2 - 3 | 484 | 9056667 | 87 | |
BC147752.1 | - | 2 - 3 | 457 | 9056701 | 87 | |
BC147753.1 | - | 2 - 3 | 457 | 9056702 | 87 | |
BC172142.1 | - | 2 - 3 | 410 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |