Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpusd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK133871.1 | - | 7 - 8 | 707 | 122 | |
BC144733.1 | - | 7 - 8 | 726 | 9053281 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |