Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159389.1 | NP_001152861.1 | 7 - 8 | 830 | 86 | |
NM_177306.3 | NP_796280.1 | 4 - 5 | 461 | 86 | ||
GenBank mRNA | AK029809.1 | - | 4 - 5 | 462 | 86 | |
AK049380.1 | - | 7 - 8 | 810 | 86 | ||
BC132104.1 | - | 2 - 3 | 144 | 40130381 | 86 | |
BC132294.1 | - | 2 - 3 | 144 | 40130571 | 86 | |
BC145065.1 | - | 2 - 3 | 144 | 86 | ||
FJ804414.1 | - | 7 - 8 | 780 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |