Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017401.2 | NP_059097.2 | 7 - 8 | 1229 | 70 | |
GenBank mRNA | AF176098.1 | - | 7 - 8 | 1002 | 70 | |
AJ251804.2 | - | 7 - 8 | 973 | 70 | ||
BC056213.1 | - | 7 - 8 | 1021 | 30024844 | 70 | |
BC056996.1 | - | 4 - 5 | 2876 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |