Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080819.1 | NP_001074288.1 | 11 - 12 | 3202 | 62 | |
GenBank mRNA | AF268912.1 | - | 10 - 11 | 2053 | 62 | |
BC082554.1 | - | 2434 | 30622048 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |