Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270435.1 | NP_001257364.1 | 8 - 9 | 1441 | 99 | |
NM_001270446.1 | NP_001257375.1 | 3 - 4 | 697 | 99 | ||
NM_028506.1 | NP_082782.1 | 4 - 5 | 704 | 99 | ||
NM_029286.2 | NP_083562.1 | 8 - 9 | 1456 | 99 | ||
GenBank mRNA | AK043853.1 | - | 3 - 4 | 690 | 99 | |
AK049010.1 | - | 3 - 4 | 597 | 99 | ||
AK078362.1 | - | 8 - 9 | 1383 | 99 | ||
AK163185.1 | - | 8 - 9 | 1382 | 99 | ||
AK224940.1 | - | 4 - 5 | 633 | 99 | ||
AK224959.1 | - | 3 - 4 | 605 | 99 | ||
AK224961.1 | - | 3 - 4 | 698 | 99 | ||
BC114436.1 | - | 8 - 9 | 1383 | 40059415 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |