Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190448.1 | NP_001177377.1 | 6 - 7 | 858 | 99 | |
NM_016672.4 | NP_057881.1 | 6 - 7 | 796 | 99 | ||
GenBank mRNA | AF071068.1 | - | 5 - 6 | 729 | 99 | |
AK149141.1 | - | 6 - 7 | 790 | 99 | ||
BC060989.1 | - | 6 - 7 | 814 | 99 | ||
BC089594.1 | - | 6 - 7 | 799 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |