Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011979.2 | NP_036109.3 | 1 - 2 | 335 | 69 | |
GenBank mRNA | AJ132103.1 | - | 1 - 2 | 325 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |