Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs3st5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253355.1 | NP_001240284.1 | 3 - 4 | 1037 | 126 | |
NM_001253356.1 | NP_001240285.1 | 2 - 3 | 978 | 126 | ||
GenBank mRNA | AK031910.1 | - | 3 - 4 | 1027 | 126 | |
AK086098.1 | - | 2 - 3 | 897 | 126 | ||
BC147490.1 | - | 3 - 4 | 1030 | 9056285 | 126 | |
BC147493.1 | - | 2 - 3 | 971 | 9056309 | 126 | |
BC150973.1 | - | 3 - 4 | 1030 | 9087886 | 126 | |
BC158004.1 | - | 3 - 4 | 1030 | 9088149 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus 1700 Probe spans exons |