Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs3st5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253355.1 | NP_001240284.1 | 4 - 5 | 1139 | 81 | |
NM_001253356.1 | NP_001240285.1 | 3 - 4 | 1080 | 81 | ||
GenBank mRNA | AK031910.1 | - | 4 - 5 | 1129 | 81 | |
BC147490.1 | - | 4 - 5 | 1132 | 9056285 | 81 | |
BC147493.1 | - | 3 - 4 | 1073 | 9056309 | 81 | |
BC150973.1 | - | 4 - 5 | 1132 | 9087886 | 81 | |
BC158004.1 | - | 4 - 5 | 1132 | 9088149 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |