Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146017.3 | NP_666129.1 | 4 - 5 | 444 | 68 | |
GenBank mRNA | AK028946.1 | - | 4 - 5 | 444 | 68 | |
AK160577.1 | - | 4 - 5 | 448 | 68 | ||
BC023693.1 | - | 4 - 5 | 431 | 5344479 | 68 | |
BC025550.1 | - | 4 - 5 | 389 | 5320737 | 68 | |
BC027245.1 | - | 4 - 5 | 407 | 3602400 | 68 | |
BC031196.1 | - | 4 - 5 | 418 | 5059564 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |