Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400027281.mm.1 |
TC0400003383.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400027281.mm.1 |
NM_001161774.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027281.mm.1 |
NM_001285520.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027281.mm.1 |
NM_001285521.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027281.mm.1 |
NM_009176.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027287.mm.1 |
TC0400003383.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400027287.mm.1 |
NM_001161774.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027287.mm.1 |
NM_001285520.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027287.mm.1 |
NM_001285521.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027287.mm.1 |
NM_009176.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027283.mm.1 |
TC0400003383.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400027283.mm.1 |
NM_001285520.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027283.mm.1 |
NM_009176.5 |
Refseq |