Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900014043.mm.1 |
NM_001166361.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014043.mm.1 |
NM_001166361.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014043.mm.1 |
NM_001166362.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014043.mm.1 |
NM_001166362.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014043.mm.1 |
TC1900001586.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900014042.mm.1 |
NM_001166361.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014042.mm.1 |
NM_001166361.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014042.mm.1 |
NM_001166362.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014042.mm.1 |
NM_001166362.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014042.mm.1 |
NM_001166363.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014042.mm.1 |
NM_001166363.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014042.mm.1 |
NM_010205.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014042.mm.1 |
NM_010205.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014042.mm.1 |
TC1900001586.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900014044.mm.1 |
NM_001166361.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014044.mm.1 |
NM_001166361.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014044.mm.1 |
TC1900001586.mm.1 |
MTA TC |