Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A1bg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081067.1 | NP_001074536.1 | 4 - 5 | 668 | 114 | |
GenBank mRNA | DQ520794.1 | - | 4 - 5 | 664 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |