Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033461.3 | NP_001028633.1 | 1 - 2 | 148 | 88 | |
NM_001289821.1 | NP_001276750.1 | 1 - 2 | 148 | 88 | ||
NM_001289822.1 | NP_001276751.1 | 1 - 2 | 148 | 88 | ||
GenBank mRNA | AK133051.1 | - | 1 - 2 | 149 | 88 | |
AK167318.1 | - | 1 - 2 | 147 | 88 | ||
AK167729.1 | - | 1 - 2 | 147 | 88 | ||
BC150919.1 | - | 1 - 2 | 162 | 9087830 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |