Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133791.4 | NP_598552.2 | 9 - 10 | 1554 | 112 | |
GenBank mRNA | AK079375.1 | - | 9 - 10 | 1389 | 112 | |
AK086956.1 | - | 9 - 10 | 1384 | 112 | ||
AK147648.1 | - | 1548 | 112 | |||
AK204107.1 | - | 1 - 2 | 35 | 112 | ||
BC067050.1 | - | 9 - 10 | 1554 | 30537299 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |