Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900010245.mm.1 |
NM_001033481.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010245.mm.1 |
NM_001033481.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010245.mm.1 |
TC1900001097.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010248.mm.1 |
NM_001033481.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010248.mm.1 |
NM_001033481.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010248.mm.1 |
TC1900001097.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010249.mm.1 |
NM_001033481.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010249.mm.1 |
NM_001033481.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010249.mm.1 |
TC1900001097.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010243.mm.1 |
NM_001033481.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010243.mm.1 |
NM_001033481.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010243.mm.1 |
TC1900001097.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010244.mm.1 |
NM_001033481.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010244.mm.1 |
NM_001033481.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010244.mm.1 |
TC1900001097.mm.1 |
MTA TC |