Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028953.2 | NP_083229.1 | 3 - 4 | 212 | 79 | |
GenBank mRNA | AF417579.1 | - | 3 - 4 | 212 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |