Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026748.2 | NP_081024.3 | 34 - 35 | 5240 | 63 | |
GenBank mRNA | AB257854.1 | - | 34 - 35 | 5200 | 63 | |
AK148183.1 | - | 5 - 6 | 698 | 63 | ||
BC010333.1 | - | 5 - 6 | 821 | 63 | ||
BC063266.1 | - | 33 - 34 | 5008 | 6409930 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |