Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168250.1 | NP_001161722.1 | 13 - 14 | 1633 | 99 | |
NM_134092.3 | NP_598853.3 | 13 - 14 | 1633 | 99 | ||
GenBank mRNA | AJ278508.1 | - | 13 - 14 | 1554 | 99 | |
AK080036.1 | - | 13 - 14 | 1447 | 99 | ||
AK088549.1 | - | 13 - 14 | 1447 | 99 | ||
AK088652.1 | - | 3 - 4 | 1061 | 99 | ||
AK133855.1 | - | 13 - 14 | 1481 | 99 | ||
BC005500.1 | - | 6 - 7 | 630 | 99 | ||
BC052197.1 | - | 13 - 14 | 1633 | 6511752 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |