Search
Search
| Gene Symbol: Ncf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286037.1 | NP_001272966.1 | 8 - 9 | 856 | 73 | |
| NM_010876.4 | NP_035006.3 | 8 - 9 | 856 | 73 | ||
| NR_104383.1 | 8 - 9 | 856 | 73 | |||
| GenBank mRNA | AB002663.1 | - | 8 - 9 | 835 | 73 | |
| AK149619.1 | - | 8 - 9 | 859 | 73 | ||
| AK149668.1 | - | 8 - 9 | 823 | 73 | ||
| AK149732.1 | - | 8 - 9 | 857 | 73 | ||
| AK150945.1 | - | 8 - 9 | 822 | 73 | ||
| AK151316.1 | - | 8 - 9 | 823 | 73 | ||
| AK151573.1 | - | 7 - 8 | 682 | 73 | ||
| AK151593.1 | - | 8 - 9 | 821 | 73 | ||
| AK152386.1 | - | 8 - 9 | 819 | 73 | ||
| AK152992.1 | - | 8 - 9 | 824 | 73 | ||
| AK162308.1 | - | 8 - 9 | 823 | 73 | ||
| AK162623.1 | - | 8 - 9 | 827 | 73 | ||
| AK170462.1 | - | 8 - 9 | 823 | 73 | ||
| AK171559.1 | - | 8 - 9 | 822 | 73 | ||
| BC026529.1 | - | 7 - 8 | 679 | 73 | ||
| BC055836.1 | - | 8 - 9 | 810 | 3983481 | 73 | |
| L11455.1 | - | 8 - 9 | 793 | 73 | ||
Set Membership: |
|