Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500010117.mm.1 |
TC0500001181.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500010117.mm.1 |
NM_138646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500010117.mm.1 |
NM_138646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500010109.mm.1 |
TC0500001181.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500010109.mm.1 |
NM_138646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500010109.mm.1 |
NM_138646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500010115.mm.1 |
TC0500001181.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500010115.mm.1 |
NM_138646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500010115.mm.1 |
NM_138646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500010116.mm.1 |
TC0500001181.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500010116.mm.1 |
NM_138646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500010116.mm.1 |
NM_138646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500010113.mm.1 |
TC0500001181.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500010113.mm.1 |
NM_138646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500010113.mm.1 |
NM_138646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500010114.mm.1 |
TC0500001181.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500010114.mm.1 |
NM_138646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500010114.mm.1 |
NM_138646.3 |
Refseq |