Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK028796.1 | - | 5 - 6 | 1266 | 70 | |
AK156499.1 | - | 4 - 5 | 1145 | 70 | ||
AK157297.1 | - | 4 - 5 | 1135 | 70 | ||
AK165124.1 | - | 3 - 4 | 374 | 70 | ||
AK171128.1 | - | 5 - 6 | 1209 | 70 | ||
AK172551.1 | - | 4 - 5 | 1135 | 70 | ||
AK172573.1 | - | 4 - 5 | 1139 | 70 | ||
BC006759.1 | - | 4 - 5 | 1119 | 3709861 | 70 | |
U89992.2 | - | 1030 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |