Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010266.2 | NP_034396.1 | 9 - 10 | 1091 | 63 | |
GenBank mRNA | AF174583.1 | - | 9 - 10 | 1017 | 63 | |
AF369880.1 | - | 9 - 10 | 976 | 63 | ||
AF369881.1 | - | 9 - 10 | 976 | 63 | ||
AF369882.1 | - | 9 - 10 | 976 | 63 | ||
AF369883.1 | - | 9 - 10 | 976 | 63 | ||
AF369884.1 | - | 9 - 10 | 976 | 63 | ||
AK041056.1 | - | 9 - 10 | 1030 | 63 | ||
AK079419.1 | - | 9 - 10 | 899 | 63 | ||
AK079459.1 | - | 9 - 10 | 1031 | 63 | ||
AK157014.1 | - | 9 - 10 | 1053 | 63 | ||
AK173138.1 | - | 9 - 10 | 1020 | 63 | ||
BC076583.1 | - | 9 - 10 | 1004 | 30620457 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |