Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnmbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY383729.1 | - | 3 - 4 | 2272 | 64 | |
CB245148.1 | - | 1 - 2 | 99 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |