Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001446.3 | NP_001001446.3 | 2 - 3 | 404 | 69 | |
NM_001167905.1 | NP_001161377.1 | 2 - 3 | 404 | 69 | ||
GenBank mRNA | AK050436.1 | - | 2 - 3 | 366 | 69 | |
AK149451.1 | - | 2 - 3 | 366 | 69 | ||
BC022141.1 | - | 2 - 3 | 68 | |||
BC024068.1 | - | 2 - 3 | 324 | 69 | ||
BC025819.1 | - | 2 - 3 | 356 | 5099385 | 69 | |
BC026766.1 | - | 2 - 3 | 356 | 4195655 | 69 | |
BC034834.1 | - | 2 - 3 | 349 | 5099935 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |