Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190795.1 | NP_001177724.1 | 3 - 4 | 313 | 118 | |
NM_026409.4 | NP_080685.2 | 3 - 4 | 313 | 118 | ||
GenBank mRNA | AK012782.1 | - | 3 - 4 | 293 | 118 | |
AK017554.1 | - | 3 - 4 | 255 | 118 | ||
AK033741.1 | - | 3 - 4 | 290 | 118 | ||
AK077525.1 | - | 3 - 4 | 294 | 118 | ||
AK129402.1 | - | 3 - 4 | 291 | 118 | ||
AK154422.1 | - | 3 - 4 | 314 | 118 | ||
BC043052.1 | - | 3 - 4 | 263 | 118 | ||
BC117786.1 | - | 3 - 4 | 275 | 40092124 | 118 | |
BC117787.1 | - | 3 - 4 | 275 | 40092125 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |