Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008861.3 | NP_032887.3 | 5 - 6 | 1494 | 75 | |
GenBank mRNA | AF014010.1 | - | 5 - 6 | 1397 | 75 | |
AK053502.1 | - | 5 - 6 | 1399 | 75 | ||
AK128961.1 | - | 6 - 7 | 1268 | 75 | ||
AK164673.1 | - | 5 - 6 | 1385 | 75 | ||
BC053058.1 | - | 5 - 6 | 1501 | 75 | ||
BC062969.1 | - | 5 - 6 | 1501 | 75 | ||
FJ609779.1 | - | 5 - 6 | 1410 | 75 | ||
Y13278.1 | - | 5 - 6 | 1410 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |