Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163575.1 | NP_001157047.1 | 7 - 8 | 1726 | 70 | |
NM_001163576.1 | NP_001157048.1 | 6 - 7 | 1830 | 70 | ||
GenBank mRNA | AK028373.1 | - | 7 - 8 | 1674 | 70 | |
AK046755.1 | - | 6 - 7 | 1811 | 70 | ||
AK153205.1 | - | 6 - 7 | 1567 | 70 | ||
BC023056.1 | - | 3 - 4 | 563 | 70 | ||
BC024074.1 | - | 7 - 8 | 1683 | 70 | ||
BC025608.1 | - | 3 - 4 | 708 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |