Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026445.2 | NP_080721.1 | 1 - 2 | 100 | 84 | |
GenBank mRNA | AK013017.1 | - | 1 - 2 | 80 | 84 | |
AK076022.1 | - | 1 - 2 | 106 | 84 | ||
AK138300.1 | - | 1 - 2 | 91 | 84 | ||
AK140169.1 | - | 1 - 2 | 71 | 84 | ||
AK157784.1 | - | 1 - 2 | 101 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons |