Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133768.4 | NP_598529.1 | 11 - 12 | 914 | 102 | |
GenBank mRNA | AK010922.1 | - | 11 - 12 | 926 | 102 | |
AK076453.1 | - | 11 - 12 | 929 | 102 | ||
AK138423.1 | - | 11 - 12 | 911 | 102 | ||
AK140160.1 | - | 11 - 12 | 929 | 102 | ||
AK148764.1 | - | 11 - 12 | 898 | 102 | ||
AK168739.1 | - | 11 - 12 | 909 | 102 | ||
BC016670.1 | - | 11 - 12 | 914 | 2648753 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |