Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008528.4 | NP_032554.2 | 4 - 5 | 684 | 80 | |
GenBank mRNA | AB015290.1 | - | 4 - 5 | 358 | 80 | |
AF068182.1 | - | 4 - 5 | 339 | 80 | ||
AJ222814.1 | - | 4 - 5 | 309 | 80 | ||
AK089704.1 | - | 4 - 5 | 324 | 80 | ||
BC059785.1 | - | 4 - 5 | 310 | 5339723 | 80 | |
Y17159.1 | - | 4 - 5 | 684 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |