Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbpms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042674.2 | NP_001036139.1 | 2 - 3 | 463 | 82 | |
NM_001042675.2 | NP_001036140.1 | 2 - 3 | 463 | 82 | ||
NM_001286168.1 | NP_001273097.1 | 2 - 3 | 463 | 82 | ||
NM_019733.3 | NP_062707.1 | 2 - 3 | 463 | 82 | ||
GenBank mRNA | AF148511.1 | - | 2 - 3 | 412 | 82 | |
AK012265.1 | - | 2 - 3 | 406 | 82 | ||
AK012586.1 | - | 2 - 3 | 406 | 82 | ||
AK041118.1 | - | 2 - 3 | 407 | 82 | ||
AK079238.1 | - | 2 - 3 | 406 | 82 | ||
BB637557.1 | - | 2 - 3 | 407 | 82 | ||
BC030397.1 | - | 2 - 3 | 424 | 4225078 | 82 | |
CK030989.1 | - | 2 - 3 | 441 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |