Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204979.1 | NP_001191908.1 | 3 - 4 | 414 | 87 | |
NM_011319.3 | NP_035449.1 | 3 - 4 | 414 | 87 | ||
GenBank mRNA | AK028358.1 | - | 3 - 4 | 346 | 87 | |
AK075827.1 | - | 3 - 4 | 345 | 87 | ||
AK077309.1 | - | 3 - 4 | 296 | 87 | ||
AK082793.1 | - | 3 - 4 | 325 | 87 | ||
AK152150.1 | - | 3 - 4 | 372 | 87 | ||
AK153158.1 | - | 3 - 4 | 364 | 87 | ||
BC008612.1 | - | 3 - 4 | 303 | 3257101 | 87 | |
BC031932.1 | - | 3 - 4 | 303 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |