Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybphl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026831.1 | NP_081107.1 | 3 - 4 | 521 | 75 | |
GenBank mRNA | AK004148.1 | - | 3 - 4 | 522 | 75 | |
BC089625.1 | - | 3 - 4 | 509 | 6756382 | 75 | |
BC132636.1 | - | 3 - 4 | 510 | 40130913 | 75 | |
BC132638.1 | - | 3 - 4 | 510 | 40130915 | 75 | |
BY704393.1 | - | 3 - 4 | 522 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |