Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033313.3 | NP_001028485.1 | 4 - 5 | 442 | 77 | |
GenBank mRNA | AK147170.1 | - | 4 - 5 | 443 | 77 | |
AK161518.1 | - | 3 - 4 | 571 | 77 | ||
AK169233.1 | - | 4 - 5 | 376 | 77 | ||
BC007162.1 | - | 3 - 4 | 342 | 77 | ||
BC055927.1 | - | 4 - 5 | 356 | 77 | ||
BC106168.1 | - | 3 - 4 | 330 | 77 | ||
BC114598.1 | - | 2 - 3 | 176 | 77 | ||
BC139204.1 | - | 4 - 5 | 395 | 8862226 | 77 | |
BC139205.1 | - | 4 - 5 | 395 | 8862227 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |