Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040699.1 | NP_001035789.1 | 6 - 7 | 797 | 82 | |
GenBank mRNA | AK028160.1 | - | 6 - 7 | 763 | 82 | |
AK081973.1 | - | 6 - 7 | 783 | 82 | ||
AK148768.1 | - | 6 - 7 | 798 | 82 | ||
BC032254.1 | - | 6 - 7 | 799 | 5368816 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |