Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040699.1 | NP_001035789.1 | 7 - 8 | 932 | 78 | |
GenBank mRNA | AK028160.1 | - | 7 - 8 | 898 | 78 | |
AK081973.1 | - | 7 - 8 | 918 | 78 | ||
AK148768.1 | - | 7 - 8 | 933 | 78 | ||
BC032254.1 | - | 7 - 8 | 934 | 5368816 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |