Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040699.1 | NP_001035789.1 | 9 - 10 | 1173 | 106 | |
GenBank mRNA | AF073882.1 | - | 2 - 3 | 232 | 106 | |
AK028160.1 | - | 9 - 10 | 1139 | 106 | ||
AK081973.1 | - | 9 - 10 | 1159 | 106 | ||
AK148768.1 | - | 9 - 10 | 1174 | 106 | ||
BC032254.1 | - | 9 - 10 | 1175 | 5368816 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |