Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oc90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010953.2 | NP_035083.1 | 4 - 5 | 265 | 81 | |
GenBank mRNA | AF091846.1 | - | 2 - 3 | 119 | 81 | |
AF093591.1 | - | 4 - 5 | 375 | 81 | ||
AK012981.1 | - | 4 - 5 | 267 | 81 | ||
BC048716.1 | - | 4 - 5 | 232 | 6334417 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |