Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdyhv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029734.1 | NP_084010.1 | 2 - 3 | 257 | 89 | |
GenBank mRNA | AK010831.1 | - | 2 - 3 | 278 | 89 | |
AK150578.1 | - | 2 - 3 | 309 | 89 | ||
BC051524.1 | - | 2 - 3 | 257 | 1514917 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |