Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdyhv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK167510.1 | - | 3 - 4 | 334 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |