Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021371.2 | NP_067346.1 | 2 - 3 | 789 | 80 | |
NM_181045.1 | NP_851388.1 | 3 - 4 | 489 | 80 | ||
GenBank mRNA | AB041539.1 | - | 2 - 3 | 780 | 80 | |
AF282251.1 | - | 3 - 4 | 471 | 80 | ||
AK035865.1 | - | 3 - 4 | 458 | 80 | ||
AK081011.1 | - | 2 - 3 | 223 | 80 | ||
AK158641.1 | - | 2 - 3 | 151 | 80 | ||
AK158892.1 | - | 2 - 3 | 761 | 80 | ||
AK163648.1 | - | 2 - 3 | 252 | 80 | ||
BC057129.1 | - | 2 - 3 | 768 | 6811387 | 80 | |
BC058963.1 | - | 2 - 3 | 768 | 6811387 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |