Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aacs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030210.1 | NP_084486.1 | 1 - 2 | 240 | 98 | |
GenBank mRNA | AB054122.1 | - | 1 - 2 | 198 | 98 | |
AB056114.1 | - | 1 - 2 | 135 | 98 | ||
AK019063.1 | - | 1 - 2 | 240 | 98 | ||
AK034184.1 | - | 1 - 2 | 173 | 98 | ||
AK043848.1 | - | 1 - 2 | 263 | 98 | ||
AK044017.1 | - | 1 - 2 | 240 | 98 | ||
AK147189.1 | - | 1 - 2 | 240 | 98 | ||
AK150266.1 | - | 1 - 2 | 240 | 98 | ||
AK165418.1 | - | 1 - 2 | 240 | 98 | ||
AK165457.1 | - | 1 - 2 | 240 | 98 | ||
AK170120.1 | - | 1 - 2 | 240 | 98 | ||
AK170653.1 | - | 1 - 2 | 240 | 98 | ||
AK170856.1 | - | 1 - 2 | 240 | 98 | ||
BC026817.1 | - | 1 - 2 | 231 | 4235112 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |