Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008146.3 | NP_032172.3 | 13 - 14 | 72 | ||
GenBank mRNA | AK133623.1 | - | 13 - 14 | 72 | ||
BC043452.1 | - | 5 - 6 | 935 | 72 | ||
BC053002.1 | - | 13 - 14 | 5709074 | 72 | ||
D78270.2 | - | 13 - 14 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |