Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sppl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029012.2 | NP_083288.2 | 10 - 11 | 1154 | 68 | |
GenBank mRNA | AK014709.1 | - | 10 - 11 | 1492 | 68 | |
BC023131.1 | - | 6 - 7 | 769 | 68 | ||
BC078437.1 | - | 1 - 2 | 1201 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |