Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008422.1 | NP_001008422.1 | 5 - 6 | 545 | 57 | |
GenBank mRNA | AK141920.1 | - | 6 - 7 | 559 | 57 | |
BC042576.1 | - | 5 - 6 | 403 | 4160122 | 57 | |
BC043138.1 | - | 5 - 6 | 575 | 6821344 | 57 | |
BC054818.1 | - | 4 - 5 | 755 | 57 | ||
BC079873.1 | - | 5 - 6 | 563 | 57 | ||
BC085153.1 | - | 5 - 6 | 545 | 6837651 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |